siRNA Details
| Gene Symbol: Tusc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030254.3 | NP_084530.3 | 445 | |||
| GenBank mRNA | AK028182.1 | - | 421 | |||
| AK088616.1 | - | 459 | ||||
| AK132766.1 | - | 468 | ||||
| AK168664.1 | - | 403 | ||||
| AK168961.1 | - | 403 | ||||
| BC003311.1 | - | 404 | ||||
| BC069898.1 | - | 445 | 6489807 | |||

Tusc3
Genomic Map