siRNA Details
| Gene Symbol: Hspb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF250139.1 | - | 1 | 343 | ||
| AF273453.1 | - | 1 | 689 | |||
| AK005052.1 | - | 1 | 711 | |||
| BC003971.1 | - | 1 | 699 | |||
| BC011219.1 | - | 1 | 692 | 4195285 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspb8| Gene Symbol: Hspb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF250139.1 | - | 1 | 343 | ||
| AF273453.1 | - | 1 | 689 | |||
| AK005052.1 | - | 1 | 711 | |||
| BC003971.1 | - | 1 | 699 | |||
| BC011219.1 | - | 1 | 692 | 4195285 | ||