siRNA Details
| Gene Symbol: Emid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080595.2 | NP_542162.1 | 3 | 232 | ||
| GenBank mRNA | AJ416093.1 | - | 3 | 232 | ||
| AK153702.1 | - | 3 | 251 | |||
| BC130023.1 | - | 3 | 237 | 40140754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emid1| Gene Symbol: Emid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080595.2 | NP_542162.1 | 3 | 232 | ||
| GenBank mRNA | AJ416093.1 | - | 3 | 232 | ||
| AK153702.1 | - | 3 | 251 | |||
| BC130023.1 | - | 3 | 237 | 40140754 | ||