siRNA Details
| Gene Symbol: Emid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080595.2 | NP_542162.1 | 15 | 1747 | ||
| GenBank mRNA | AJ416093.1 | - | 15 | 1747 | ||
| AK153702.1 | - | 17 | 1925 | |||
| BC012248.1 | - | 6 | 851 | |||
| BC021849.1 | - | 5 | 753 | |||

Emid1
Genomic Map