siRNA Details
Gene Symbol: Sertad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133210.2 | NP_573473.1 | 2 | 1137 | ||
GenBank mRNA | AK134829.1 | - | 2 | 1210 | ||
BC034886.1 | - | 2 | 1137 | 1364881 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sertad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133210.2 | NP_573473.1 | 2 | 1137 | ||
GenBank mRNA | AK134829.1 | - | 2 | 1210 | ||
BC034886.1 | - | 2 | 1137 | 1364881 |