siRNA Details
| Gene Symbol: Tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133683.3 | NP_598444.1 | 5 | 735 | ||
| GenBank mRNA | AK018383.1 | - | 5 | 683 | ||
| AK034298.1 | - | 5 | 741 | |||
| AK036228.1 | - | 4 | 812 | |||
| AK042426.1 | - | 4 | 790 | |||
| AK079735.1 | - | 593 | ||||
| AK136778.1 | - | 724 | ||||
| AK138931.1 | - | 5 | 723 | |||
| AK155275.1 | - | 5 | 696 | |||
| AK170143.1 | - | 5 | 695 | |||
| AK171087.1 | - | 5 | 695 | |||
| BC016895.1 | - | 5 | 737 | 4239007 | ||

Tmem19
Genomic Map