siRNA Details
| Gene Symbol: Gmppa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133708.3 | NP_598469.1 | 13 | 1322 | ||
| GenBank mRNA | AK007463.1 | - | 13 | 1307 | ||
| AK144311.1 | - | 13, 14 | 1587 | |||
| AK150473.1 | - | 14 | 1458 | |||
| AK209171.1 | - | 1 | 41 | |||
| BC008116.1 | - | 13 | 1281 | 3498021 | ||

Gmppa
Genomic Map