siRNA Details
| Gene Symbol: Chrdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291320.1 | NP_001278249.1 | 9, 10 | 1236 | ||
| NM_133709.3 | NP_598470.3 | 9, 10 | 1278 | |||
| GenBank mRNA | AF338222.1 | - | 9, 10 | 1182 | ||
| AK007577.1 | - | 9, 10 | 1241 | |||
| BC019399.1 | - | 9, 10 | 1243 | 4012143 | ||

Chrdl2
Genomic Map