siRNA Details
| Gene Symbol: Lancl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133737.2 | NP_598498.1 | 1, 2 | 805 | ||
| GenBank mRNA | AB035967.1 | - | 1, 2 | 624 | ||
| AK005632.1 | - | 1, 2 | 174 | |||
| BC016072.1 | - | 1, 2 | 659 | 4482042 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lancl2| Gene Symbol: Lancl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133737.2 | NP_598498.1 | 1, 2 | 805 | ||
| GenBank mRNA | AB035967.1 | - | 1, 2 | 624 | ||
| AK005632.1 | - | 1, 2 | 174 | |||
| BC016072.1 | - | 1, 2 | 659 | 4482042 | ||