siRNA Details
| Gene Symbol: Mmaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133823.4 | NP_598584.2 | 7 | 2652 | ||
| GenBank mRNA | AK050255.1 | - | 7 | 2633 | ||
| BC085229.1 | - | 6 | 2611 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmaa| Gene Symbol: Mmaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133823.4 | NP_598584.2 | 7 | 2652 | ||
| GenBank mRNA | AK050255.1 | - | 7 | 2633 | ||
| BC085229.1 | - | 6 | 2611 | |||