siRNA Details
| Gene Symbol: Rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144528.3 | NP_653111.1 | 9 | 1334 | ||
| GenBank mRNA | AK011369.1 | - | 1 | 211 | ||
| AK042319.1 | - | 8 | 1131 | |||
| AK154781.1 | - | 9 | 1300 | |||
| BB823711.1 | - | 1 | 124 | |||
| BC016543.1 | - | 9 | 1306 | 3158083 | ||
| CF582846.1 | - | 1 | 342 | |||

Rnf126
Genomic Map