siRNA Details
| Gene Symbol: Kdm4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172095.1 | NP_001165566.1 | 6 | 826 | ||
| NM_144787.2 | NP_659036.1 | 6 | 890 | |||
| GenBank mRNA | AK042792.1 | - | 5 | 605 | ||
| AK048092.1 | - | 6 | 803 | |||
| AK053104.1 | - | 6 | 766 | |||
| AK081769.1 | - | 6 | 833 | |||
| AK144103.1 | - | 6 | 853 | |||
| AK144890.1 | - | 6 | 864 | |||
| AK173023.1 | - | 6 | 890 | |||
| BC020180.1 | - | 6 | 832 | 4923943 | ||
| BC042424.1 | - | 6 | 826 | 5357211 | ||

Kdm4c
Genomic Map