siRNA Details
| Gene Symbol: Kdm4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172095.1 | NP_001165566.1 | 11 | 1684 | ||
| NM_144787.2 | NP_659036.1 | 11 | 1748 | |||
| GenBank mRNA | AK053104.1 | - | 11 | 1624 | ||
| AK081769.1 | - | 11 | 1691 | |||
| AK144103.1 | - | 11 | 1711 | |||
| AK144890.1 | - | 11 | 1722 | |||
| AK173023.1 | - | 11 | 1748 | |||
| BC020180.1 | - | 11 | 1690 | 4923943 | ||
| BC042424.1 | - | 11 | 1684 | 5357211 | ||

Kdm4c
Genomic Map