siRNA Details
| Gene Symbol: Senp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144851.5 | NP_659100.1 | 10 | 1344 | ||
| GenBank mRNA | AK028565.1 | - | 10 | 1086 | ||
| AK028762.1 | - | 2 | 2318 | |||
| AK035581.1 | - | 10 | 1180 | |||
| AK035723.1 | - | 11 | 1527 | |||
| AK054521.1 | - | 11 | 1476 | |||
| AK088597.1 | - | 10 | 1088 | |||
| BC023129.1 | - | 9 | 1058 | 3595534 | ||

Senp1
Genomic Map