siRNA Details
| Gene Symbol: Msto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144898.2 | NP_659147.2 | 2 | 267 | ||
| GenBank mRNA | AK139708.1 | - | 250 | |||
| AK163280.1 | - | 2 | 127 | |||
| AK166236.1 | - | 2 | 239 | |||
| AK166834.1 | - | 1 | 311 | |||
| AK167586.1 | - | 2 | 215 | |||
| AK170160.1 | - | 2 | 250 | |||
| BC008103.1 | - | 2 | 217 | 3966815 | ||
| BC108354.1 | - | 2 | 201 | 5035560 | ||
| BY267786.1 | - | 2 | 268 | |||

Msto1
Genomic Map