siRNA Details
| Gene Symbol: Msto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144898.2 | NP_659147.2 | 6 | 567 | ||
| GenBank mRNA | AK134001.1 | - | 24 | |||
| AK139708.1 | - | 546 | ||||
| AK163280.1 | - | 6 | 427 | |||
| AK166236.1 | - | 6 | 539 | |||
| AK166834.1 | - | 1 | 1855 | |||
| AK167586.1 | - | 6 | 515 | |||
| AK170160.1 | - | 6 | 550 | |||
| AK186611.1 | - | 2 | 120 | |||
| BC008103.1 | - | 6 | 517 | 3966815 | ||
| BC108354.1 | - | 6 | 501 | 5035560 | ||

Msto1
Genomic Map