siRNA Details
| Gene Symbol: Rnf144b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170643.1 | NP_001164114.1 | 8 | 2923 | ||
| NM_146042.4 | NP_666154.3 | 8 | 2959 | |||
| GenBank mRNA | AK030066.1 | - | 1 | 1257 | ||
| AK040939.1 | - | 8 | 2915 | |||
| AK052846.1 | - | 8 | 2924 | |||
| AK053529.1 | - | 8 | 2959 | |||
| BC025007.1 | - | 8 | 2903 | 5050848 | ||

Rnf144b
Genomic Map