siRNA Details
| Gene Symbol: Zfp553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146201.1 | NP_666313.1 | 2 | 2460 | ||
| GenBank mRNA | AK146581.1 | - | 2 | 2680 | ||
| AK148378.1 | - | 2 | 2611 | |||
| BC012403.1 | - | 1 | 2321 | |||
| BC026401.1 | - | 2 | 2460 | 4209324 | ||

Zfp553
Genomic Map