siRNA Details
| Gene Symbol: Plcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152813.3 | NP_690026.2 | 6 | 1042 | ||
| GenBank mRNA | AK046669.1 | - | 6 | 1008 | ||
| AK164809.1 | - | 5 | 1284 | |||
| AK166977.1 | - | 4 | 2383 | |||
| AK173311.1 | - | 5 | 1290 | |||
| BC031392.1 | - | 6 | 986 | 4952407 | ||

Plcd3
Genomic Map