siRNA Details
| Gene Symbol: Sec16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153125.2 | NP_694765.2 | 30 | 8374 | ||
| GenBank mRNA | AK122242.2 | - | 23 | 3811 | ||
| BC025624.1 | - | 6 | 1626 | |||
| BC034649.1 | - | 21 | 3492 | |||
| BC042603.1 | - | 25 | 3949 | |||
| BC059898.1 | - | 21 | 3978 | |||

Sec16a
Genomic Map