siRNA Details
| Gene Symbol: Ankrd23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153502.3 | NP_705722.2 | 3 | 273 | ||
| GenBank mRNA | AF492400.1 | - | 3 | 273 | ||
| BC022973.1 | - | 432 | ||||
| BC023221.1 | - | 3 | 250 | |||
| BC132370.1 | - | 3 | 283 | 40130647 | ||
| BC138797.1 | - | 3 | 283 | 8861819 | ||
| BC145421.1 | - | 3 | 283 | 9053979 | ||

Ankrd23
Genomic Map