siRNA Details
| Gene Symbol: Slc16a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172371.3 | NP_758959.1 | 4 | 2331 | ||
| GenBank mRNA | AK005699.1 | - | 3 | 1757 | ||
| AK028143.1 | - | 4 | 2316 | |||
| AK028759.1 | - | 4 | 2332 | |||
| BC039949.1 | - | 4 | 2119 | 4167719 | ||

Slc16a13
Genomic Map