siRNA Details
| Gene Symbol: Dtx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172442.3 | NP_766030.3 | 2 | 1487 | ||
| GenBank mRNA | AK086624.1 | - | 1 | 55 | ||
| BC058647.1 | - | 2 | 945 | 6405881 | ||
| BC069975.1 | - | 1 | 27 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtx4| Gene Symbol: Dtx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172442.3 | NP_766030.3 | 2 | 1487 | ||
| GenBank mRNA | AK086624.1 | - | 1 | 55 | ||
| BC058647.1 | - | 2 | 945 | 6405881 | ||
| BC069975.1 | - | 1 | 27 | |||