siRNA Details
| Gene Symbol: Nek11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172461.3 | NP_766049.2 | 7 | 1014 | ||
| GenBank mRNA | AK030042.1 | - | 7 | 973 | ||
| AK030186.1 | - | 6 | 890 | |||
| AK043543.1 | - | 7 | 1253 | |||
| AK054237.1 | - | 7 | 908 | |||
| BC119588.1 | - | 6 | 848 | 40058889 | ||
| BC119589.1 | - | 6 | 848 | 40058890 | ||

Nek11
Genomic Map