siRNA Details
| Gene Symbol: AP1M2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300887.1 | NP_001287816.1 | 4, 5 | 488 | ||
| NM_005498.4 | NP_005489.2 | 4, 5 | 488 | |||
| GenBank mRNA | AF020797.2 | - | 4, 5 | 451 | ||
| AK023863.1 | - | 4, 5 | 489 | |||
| AK222946.1 | - | 4, 5 | 454 | |||
| AK315689.1 | - | 4, 5 | 479 | |||
| BC003387.2 | - | 4, 5 | 426 | 3446133 | ||
| BC003612.2 | - | 4, 5 | 425 | 2988458 | ||
| BC005021.1 | - | 4, 5 | 488 | 3139433 | ||

AP1M2
Genomic Map