siRNA Details
| Gene Symbol: Astl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291003.1 | NP_001277932.1 | 9 | 1407 | ||
| NM_172539.3 | NP_766127.1 | 9 | 1538 | |||
| GenBank mRNA | AK033037.1 | - | 9 | 1538 | ||
| BC064729.1 | - | 9 | 1399 | 30476764 | ||

Astl
Genomic Map