siRNA Details
| Gene Symbol: Tmem120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172541.2 | NP_766129.1 | 3, 4 | 367 | ||
| GenBank mRNA | AK028119.1 | - | 3, 4 | 368 | ||
| AK217162.1 | - | 2, 3 | 181 | |||
| BC046757.1 | - | 3, 4 | 327 | 5693443 | ||

Tmem120a
Genomic Map