siRNA Details
| Gene Symbol: Klhl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172565.2 | NP_766153.1 | 1 | 531 | ||
| GenBank mRNA | AK029101.1 | - | 1 | 532 | ||
| AK169910.1 | - | 2 | 429 | |||
| BC111873.1 | - | 1 | 527 | 40059437 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl11| Gene Symbol: Klhl11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172565.2 | NP_766153.1 | 1 | 531 | ||
| GenBank mRNA | AK029101.1 | - | 1 | 532 | ||
| AK169910.1 | - | 2 | 429 | |||
| BC111873.1 | - | 1 | 527 | 40059437 | ||