siRNA Details
| Gene Symbol: Zfp322a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111107.2 | NP_001104577.1 | 5 | 2622 | ||
| NM_001286155.1 | NP_001273084.1 | 5 | 2900 | |||
| NM_001286156.1 | NP_001273085.1 | 4 | 2825 | |||
| NM_172586.4 | NP_766174.1 | 4 | 2547 | |||
| GenBank mRNA | AK036877.1 | - | 4 | 2548 | ||
| AK162608.1 | - | 4 | 2525 | |||
| AK170525.1 | - | 4 | 2542 | |||
| BC043711.1 | - | 4 | 2509 | 5369339 | ||

Zfp322a
Genomic Map