siRNA Details
| Gene Symbol: Ttc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172770.3 | NP_766358.1 | 6, 7 | 544 | ||
| GenBank mRNA | AK054350.1 | - | 6, 7 | 545 | ||
| AK076457.1 | - | 5, 6 | 1035 | |||
| BC056616.1 | - | 5, 6 | 472 | 4974294 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc12| Gene Symbol: Ttc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172770.3 | NP_766358.1 | 6, 7 | 544 | ||
| GenBank mRNA | AK054350.1 | - | 6, 7 | 545 | ||
| AK076457.1 | - | 5, 6 | 1035 | |||
| BC056616.1 | - | 5, 6 | 472 | 4974294 | ||