siRNA Details
Gene Symbol: Ttc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172770.3 | NP_766358.1 | 22 | 3532 | ||
GenBank mRNA | AK054350.1 | - | 22 | 3533 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ttc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172770.3 | NP_766358.1 | 22 | 3532 | ||
GenBank mRNA | AK054350.1 | - | 22 | 3533 |