siRNA Details
| Gene Symbol: MLANA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005511.1 | NP_005502.1 | 5 | 780 | ||
| GenBank mRNA | U06452.1 | - | 5 | 780 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLANA| Gene Symbol: MLANA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005511.1 | NP_005502.1 | 5 | 780 | ||
| GenBank mRNA | U06452.1 | - | 5 | 780 | ||