siRNA Details
| Gene Symbol: Slco5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172841.2 | NP_766429.2 | 1 | 860 | ||
| GenBank mRNA | AK028758.1 | - | 2 | 762 | ||
| AK041736.1 | - | 1 | 861 | |||
| AK048882.1 | - | 1 | 785 | |||
| BC117943.1 | - | 1 | 639 | 40097907 | ||
| BC117944.1 | - | 1 | 639 | |||

Slco5a1
Genomic Map