siRNA Details
| Gene Symbol: Snx14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172926.3 | NP_766514.2 | 7 | 744 | ||
| GenBank mRNA | AK046839.1 | - | 7 | 686 | ||
| AK049371.1 | - | 7 | 723 | |||
| BC080799.1 | - | 7 | 729 | 4948538 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx14| Gene Symbol: Snx14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172926.3 | NP_766514.2 | 7 | 744 | ||
| GenBank mRNA | AK046839.1 | - | 7 | 686 | ||
| AK049371.1 | - | 7 | 723 | |||
| BC080799.1 | - | 7 | 729 | 4948538 | ||