siRNA Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172993.3 | NP_766581.2 | 3 | 325 | ||
| GenBank mRNA | AK084205.1 | - | 3 | 272 | ||
| AK146353.1 | - | 3 | 289 | |||
| AK149921.1 | - | 3 | 272 | |||
| AK154918.1 | - | 3 | 272 | |||
| AK173267.1 | - | 3 | 300 | |||
| BC021520.1 | - | 1 | 135 | |||
| BC066136.1 | - | 3 | 248 | |||
| BC089160.1 | - | 3 | 247 | 5390706 | ||
| BY125394.1 | - | 2 | 267 | |||

Zfp512
Genomic Map