siRNA Details
| Gene Symbol: Zfp512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172993.3 | NP_766581.2 | 6, 7 | 711 | ||
| GenBank mRNA | AK084205.1 | - | 6, 7 | 658 | ||
| AK146353.1 | - | 6, 7 | 675 | |||
| AK149921.1 | - | 6, 7 | 658 | |||
| AK154918.1 | - | 6, 7 | 658 | |||
| AK173267.1 | - | 6, 7 | 686 | |||
| BC021520.1 | - | 4, 5 | 521 | |||
| BC066136.1 | - | 6, 7 | 634 | |||
| BC089160.1 | - | 6, 7 | 633 | 5390706 | ||

Zfp512
Genomic Map