siRNA Details
| Gene Symbol: Usp43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291049.1 | NP_001277978.1 | 7, 8 | 1354 | ||
| NM_173754.4 | NP_776115.2 | 7, 8 | 1354 | |||
| GenBank mRNA | AK042217.1 | - | 7, 8 | 1355 | ||
| AK083271.1 | - | 7, 8 | 1192 | |||
| BC021474.1 | - | 6, 7 | 644 | 5343429 | ||

Usp43
Genomic Map