siRNA Details
| Gene Symbol: Mrps27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173757.3 | NP_776118.2 | 11 | 1375 | ||
| GenBank mRNA | AK076008.1 | - | 11 | 1374 | ||
| AK087686.1 | - | 11 | 1374 | |||
| AK172922.1 | - | 3 | 5167 | |||
| BC049149.1 | - | 11 | 1302 | 6309524 | ||

Mrps27
Genomic Map