siRNA Details
| Gene Symbol: Mrps27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173757.3 | NP_776118.2 | 11 | 1542 | ||
| GenBank mRNA | AA461997.1 | - | 1 | 31 | ||
| AK076008.1 | - | 11 | 1541 | |||
| AK087686.1 | - | 11 | 1542 | |||
| AK172922.1 | - | 3 | 5334 | |||
| AK185458.1 | - | 1 | 231 | |||
| BC049149.1 | - | 11 | 1469 | 6309524 | ||

Mrps27
Genomic Map