siRNA Details
| Gene Symbol: Cenpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173762.4 | NP_776123.3 | 4 | 427 | ||
| GenBank mRNA | AB001426.1 | - | 1 | 88 | ||
| AK133445.1 | - | 4 | 428 | |||
| AY493378.1 | - | 4 | 331 | |||
| BC039755.1 | - | 4 | 362 | |||
| BC052843.1 | - | 4 | 369 | |||
| BC080703.1 | - | 4 | 373 | |||
| BC106096.1 | - | 4 | 340 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_006233350, XM_006233351, XM_006233352.

Cenpe
Genomic Map