siRNA Details
| Gene Symbol: Mamdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174857.3 | NP_777282.1 | 4, 5 | 762 | ||
| GenBank mRNA | AK004794.1 | - | 4, 5 | 882 | ||
| AK035566.1 | - | 4, 5 | 763 | |||
| AK154219.1 | - | 4, 5 | 839 | |||
| BC042773.1 | - | 4, 5 | 791 | 4019075 | ||

Mamdc2
Genomic Map