siRNA Details
| Gene Symbol: Plekhf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175175.4 | NP_780384.1 | 2 | 2802 | ||
| GenBank mRNA | AK004426.1 | - | 1 | 416 | ||
| AK141254.1 | - | 1 | 1382 | |||
| BC016134.1 | - | 2 | 2743 | 4503757 | ||
| BC039276.1 | - | 2 | 2746 | 4987735 | ||

Plekhf2
Genomic Map