siRNA Details
| Gene Symbol: Golim4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291069.1 | NP_001277998.1 | 9 | 1598 | ||
| NM_175193.6 | NP_780402.1 | 8 | 1514 | |||
| GenBank mRNA | AK033730.1 | - | 8 | 1501 | ||
| AK048434.1 | - | 9 | 1425 | |||
| AK078928.1 | - | 9 | 1598 | |||
| BC086642.1 | - | 8 | 1346 | 6836803 | ||
| BC089354.1 | - | 8 | 1396 | 6837211 | ||

Golim4
Genomic Map