siRNA Details
| Gene Symbol: Arid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175251.4 | NP_780460.3 | 15 | 3013 | ||
| GenBank mRNA | AK014481.1 | - | 2 | 978 | ||
| AK173201.1 | - | 1 | 840 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arid2| Gene Symbol: Arid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175251.4 | NP_780460.3 | 15 | 3013 | ||
| GenBank mRNA | AK014481.1 | - | 2 | 978 | ||
| AK173201.1 | - | 1 | 840 | |||