siRNA Details
| Gene Symbol: Arid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175251.4 | NP_780460.3 | 21 | 5803 | ||
| GenBank mRNA | AK014481.1 | - | 8 | 3768 | ||
| AK034964.1 | - | 7 | 1964 | |||
| AK036304.1 | - | 2 | 382 | |||
| AK043864.1 | - | 1 | 632 | |||
| AK077022.1 | - | 5 | 2042 | |||
| AK077366.1 | - | 7 | 2565 | |||
| AK173201.1 | - | 7 | 3630 | |||
| BC027180.1 | - | 7 | 2702 | 4021271 | ||

Arid2
Genomic Map