siRNA Details
| Gene Symbol: Zfp526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175436.4 | NP_780645.1 | 3 | 2485 | ||
| GenBank mRNA | AK050839.1 | - | 3 | 2486 | ||
| AK051798.1 | - | 3 | 2483 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp526| Gene Symbol: Zfp526 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175436.4 | NP_780645.1 | 3 | 2485 | ||
| GenBank mRNA | AK050839.1 | - | 3 | 2486 | ||
| AK051798.1 | - | 3 | 2483 | |||