siRNA Details
| Gene Symbol: Slc35g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175507.3 | NP_780716.1 | 3 | 759 | ||
| GenBank mRNA | AK041618.1 | - | 3 | 759 | ||
| AK052365.1 | - | 3 | 663 | |||
| AK171987.1 | - | 3 | 691 | |||
| AK209992.1 | - | 1 | 107 | |||
| BC137663.1 | - | 3 | 770 | 8860682 | ||
| BC137664.1 | - | 3 | 770 | 8860683 | ||

Slc35g1
Genomic Map