siRNA Details
| Gene Symbol: Fbxo46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175530.3 | NP_780739.1 | 2 | 2761 | ||
| GenBank mRNA | AK030147.1 | - | 2 | 2749 | ||
| BC052088.1 | - | 1 | 983 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo46| Gene Symbol: Fbxo46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175530.3 | NP_780739.1 | 2 | 2761 | ||
| GenBank mRNA | AK030147.1 | - | 2 | 2749 | ||
| BC052088.1 | - | 1 | 983 | |||