siRNA Details
| Gene Symbol: Wdfy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175546.4 | NP_780755.2 | 5 | 583 | ||
| GenBank mRNA | AK045072.1 | - | 4 | 314 | ||
| AK086275.1 | - | 5 | 596 | |||
| AK131958.1 | - | 5 | 582 | |||
| AK151817.1 | - | 5 | 539 | |||
| BC116632.1 | - | 5 | 529 | 40056912 | ||

Wdfy2
Genomic Map