siRNA Details
| Gene Symbol: Hopx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159900.1 | NP_001153372.1 | 3 | 662 | ||
| NM_001159901.1 | NP_001153373.1 | 2 | 790 | |||
| NM_175606.3 | NP_783199.1 | 3 | 822 | |||
| GenBank mRNA | AF492703.1 | - | 3 | 817 | ||
| AF492704.1 | - | 3 | 645 | |||
| AK009007.1 | - | 3 | 579 | |||

Hopx
Genomic Map